DuPont Molykote High-Vacuum Grease
DuPont Molykote High-Vacuum Grease
DuPont Molykote High-Vacuum Grease
Have Questions?
Catalog NumberQuantity
FKS154719771 Ea.
Catalog number FKS15471977
Price (SGD)
324.00
EA
Add to cart
Quantity:
1 Ea.
Request bulk or custom format
Price (SGD)
324.00
EA
Add to cart
Specifications
For Use With (Application)Used as an anti-seize grease in ground glass joints. Also used to lubricate synthetic and silicone rubber gaskets. Further, it is used to lubricate plug valves, control valves, flow meter bearings, ceramic plug cocks and fire extinguisher valves. In addition to this, it is used to lubricate water-treating equipment, laboratory equipment, lubricate O-rings on binoculars and telescopes and to prevent fogging of delicate lenses.
PackagingTube
Quantity1 Ea.
Temperature (Metric)-40°C to +204°C
Weight (English)5.3 oz.
View more
Recommended for vacuums reaching between 1 x 10-5 and 1 x 10-6 mm HgDow Corning™ High-Vacuum Grease is used as an anti-seize grease in ground glass joints. It is used to lubricate synthetic and silicone rubber gaskets. Further, it is used to lubricate plug valves, control valves, flow meter bearings, ceramic plug cocks and fire extinguisher valves. In addition to this, it is used to lubricate water-treating equipment, laboratory equipment, lubricate O-rings on binoculars and telescopes and to prevent fogging of delicate lenses.