Borosilicate Glass Tall Form Beakers
Actual product may vary
Borosilicate Glass Tall Form Beakers
Includes graduations and a spout for easy pouring. Fisherbrand™ Borosilicate Glass Tall Form Beakers are available in a variety of sizes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS154690931000 mL
FKS15449093600 mL
Catalog number FKS15469093
Price (SGD)
162.00
Pack
Capacity (Metric):
1000 mL
Price (SGD)
162.00
Pack
  • Borosilicate Glass 3.3
  • Tall Form Graduated with Spout
  • Manufactured to ISO 3819
Specifications
ColorTranslucent
Diameter (Metric) Outer95 mm
Capacity (Metric)1000 mL
MaterialBorosilicate Glass (3.3)
FinishMatte
Height (Metric)180 mm
IncludesSpout
Quantity10 Pk.
TypeBeaker
AutoclavableAutoclavable
DisposableReusable
GraduatedYes
Certifications/ComplianceISO 3819
ScaleWhite Graduated Scale
StyleTall Form
Unit SizePack