Dumont Self-Closing #5 Tweezers
Product Image
Fisher Catalogue
Dumont Self-Closing #5 Tweezers
A pair of straight and self-closing tweezers perfectly suited for biological applications in the laboratory.
Have Questions?
Catalog number FKS15443582
View Price:Sign in to see your account pricing. Need an account? Register with us today.
  • 11cm long
  • 0.05 x 0.01mm tips for very fine work
  • Manufactured from stainless steel
  • Autoclavable with general corrosion resistance
  • Hardness measurement of 55-56, which makes them strong enough to resist breakage but flexible enough to bend slightly
  • Maximum temperature resistance of 350°C
Specifications
MaterialStainless Steel
Length (Metric)11 cm
Product TypeTweezers
For Use With (Application)General applications, biology
Thickness (Metric) Tip0.05 x 0.01mm
StyleSelf-closing Straight
Unit SizeEA