Microcentrifuge Tubes with Locking Snap Cap
Microcentrifuge Tubes with Locking Snap Cap
Actual product may vary
Microcentrifuge Tubes with Locking Snap Cap
Microcentrifuge Tubes with Locking Snap Cap
Fisherbrand
Microcentrifuge Tubes with Locking Snap Cap
Polypropylene microcentrifuge tubes are DNase/RNase and pyrogen free
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorSterilityCapacity (Metric)Max. RCF
FKS15442545NaturalNon-sterile2.0 mL25,000 x g
FIS14-666-313NaturalSterile2.0 mL25,000 x g
FIS14-666-315NaturalNon-sterile2.0 mL25,000 x g
FIS14-666-318NaturalSterile1.5 mL26,000 x g
FIS14-666-319NaturalNon-sterile1.5 mL26,000 x g
FKS15432545NaturalNon-sterile1.5 mL26,000 x g
FIS14-666-323RedNon-sterile1.5 mL26,000 x g
FIS14-666-332NaturalSterile0.5 mL30,000 x g
Catalog number FKS15442545
Price (SGD)
43.00
PK
Add to cart
Color:
Natural
Sterility:
Non-sterile
Capacity (Metric):
2.0 mL
Max. RCF:
25,000 x g
Price (SGD)
43.00
PK
Add to cart
Polypropylene microcentrifuge tubes are DNase/RNase and pyrogen free
  • DNase/RNase and pyrogen free
  • Available in both Sterile and Nonsterile
  • Autoclavable and withstand temperatures to -86°C
  • Graduated tubes available in three sizes - 0.5mL, 1.5mL and 2.0mL
Specifications
Cap StyleLocking snap cap
AutoclavableAutoclavable
Capacity (Metric)2.0 mL
ColorNatural
Length (Metric)40 mm
MaterialPolypropylene
SterilityNon-sterile
GraduatedYes
Max. RCF25,000 x g
Diameter (Metric)13 mm
Unit SizePK