SP Scienceware™ 144-Place PCR Tube Freezer Storage Boxes
SP Scienceware™ 144-Place PCR Tube Freezer Storage Boxes
Actual product may vary
Fisherbrand
SP Scienceware™ 144-Place PCR Tube Freezer Storage Boxes
Provide ULT freezer storage at temperatures down to -86°C
Have Questions?
Catalog NumberDimensions (L x W x H)
FKS154277105.25 x 5.25 x 1 in.
Catalog number FKS15427710
Price (SGD)
219.00
PK
Add to cart
Dimensions (L x W x H):
5.25 x 5.25 x 1 in.
Price (SGD)
219.00
PK
Add to cart
Provide ULT freezer storage at temperatures down to -86°C
Designed to fit in a standard 2 in. freezer rack, these 144-place PCR Tube Freezer Boxes will hold individual and/or strip 0.2ml PCR tubes.
  • Two boxes may be stacked to fit together in one space in a 2 in. freezer storage rack if using flat-capped PCR tubes
  • Clear polycarbonate lid and base and alpha-numeric index make it easy to identify samples within the box
  • Hold individual and/or strip 0.2mL PCR tubes
  • Durable and impervious to moisture
  • L x W x H: 13.2 x 13.2 x 2.5cm (5.25 x 5.25 x 1 in.)
Specifications
Closure TypeLid
ColorClear
MaterialPolycarbonate
ShapeSquare
For Use With (Equipment)2 in. freezer storage rack
Holds0.2 mL PCR Tubes
Product TypeFreezer Box
Quantity5/Pk.
Dimensions (L x W x H)5.25 x 5.25 x 1 in.
Length (Metric)13.2 cm
Length (English)5.2 in.
Width (Metric)13.2 cm
Width (English)5.2 in.
Height (Metric)2.5 cm
Height (English)1 in.
Unit SizePK