Stackable Industrial Measuring Jug
Stackable Industrial Measuring Jug
Actual product may vary
Stackable Industrial Measuring Jug
Light and handy Polypropylene measuring jug suitable for industrial applications. Includes two measuring scales, embossed transparent and with blue lettering. With ergonomic handle, available in closed design for special stability or with open handle for stacking.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS154057642000 mL
FKS152299783000 mL
Catalog number FKS15405764
Price (SGD)
47.00
EA
Add to cart
Capacity (Metric):
2000 mL
Price (SGD)
47.00
EA
Add to cart
  • Suitable for industrial applications.
  • Includes two embossed measuring scales:
    • Transparent and with blue lettering.
  • Ergonomic handle, available in closed design for special stability or with open handle for stacking to save space.
  • Polypropylene construction and highly resistant to acids, bases and chemicals.
  • Transparent
Specifications
Capacity (Metric)2000 mL
ColorTransparent
For Use With (Application)Foodstuffs
MaterialPolypropylene
Weight (Metric)0.181 kg
Graduations20 mL
GraduatedYes
Height (Metric)206 mm
Diameter (Metric)151 mm
Unit SizeEA