Search Thermo Fisher Scientific
Catalog Number | Volume (Metric) | Sterility | Bulb Draw (Metric) | Packaging Type |
---|---|---|---|---|
FKS15397883 | 4.9 mL | Sterile | 1.8 mL | Individually Wrapped 500/Pk |
FIS13-711-21 | 7.7 mL | Sterile | 3.5 mL | Individually Wrapped 500/Pk |
FIS13-711-23 | 8.6 mL | Non-sterile | 6.0 mL | Bulk 400/Pk |
FKS15327863 | 1.8 mL | Sterile | 0.9 mL | 20/Bag, 500/Pk |
336 also known as HC3NMPAFLAB36 | 4.9 mL | Non-sterile | 1.8 mL | Bulk 500/Pk |
FKS15317883 | 3.1 mL | Non-sterile | 2.2 mL | Bulk, 400/Pk. |
Recommended for: Research Applications