Klercide 70/30 IPA Sterile 6x1L Spray
Klercide 70/30 IPA Sterile 6x1L Spray
Actual product may vary
Fisherbrand
Klercide 70/30 IPA Sterile 6x1L Spray
Ensures a spore free alcohol by irradiating at no less than 25 kGy. Ecolab™ Klercide 70/30 IPA Steril 6x1L Spray preserves sterility of the contents with the patented SteriShield Delivery System and an adjustable trigger spray.
Have Questions?
Catalog NumberVolume (Metric)
FKS153960781 L
Catalog number FKS15396078
Price (SGD)
1,048.00
PK
Add to cart
Volume (Metric):
1 L
Price (SGD)
1,048.00
PK
Add to cart
Formerly Premier Klercide 70/30 Sterile IPA.Sterile spore free 70/30 v/v alcohol, either IPA or Denatured Ethanol blended with deionised water.
  • 0.2 micron filtered alcohol blend, filled and double bagged in a Grade C (ISO class 7) cleanroom, before irradiation using a validated process at no less than 25 kGy to ensure a spore free alcohol.
  • 70% isopropyl alcohol blended with deionized water.
  • Supplied sterile by gamma irradiation and double bagged.
  • Suggested Usage
    • Grade A: Very Suitable
    • Grade B: Very Suitable
    • Grade C/D: Suitable
    • Unclassified: Suitable
  • Efficacy
    • Bactericida: EN 1276
    • Fungicidal: EN 1650
    • Bactericidal and Fungicidal: EN 13697
Specifications
Product TypeDisinfectant
Composition70% isopropyl alcohol blended with deionized water
For Use With (Application)Disinfecting controlled environmnt
Quantity6/Pk.
Shelf Life3 Months
Volume (Metric)1 L
Unit SizePK