1x1 image pixel for data collection
myFuge™ 12 Microcentrifuge with COMBI-Rotor™
myFuge™ 12 Microcentrifuge with COMBI-Rotor™
Actual product may vary
Fisherbrand
myFuge™ 12 Microcentrifuge with COMBI-Rotor™
The myFuge™ 12 eliminates the need to change rotors when switching between microtubes and PCR strips. The included COMBI-Rotor™ accommodates 12 microtubes and 4 PCR strips, all at the same time.
Have Questions?
Catalog NumberElectrical Requirements
FKS15386837100/240 V 50/60 Hz
Catalog number FKS15386837
Price (SGD)
1,963.00
EA
Estimated availability date 01-Oct-2025
Add to cart
Electrical Requirements:
100/240 V 50/60 Hz
Price (SGD)
1,963.00
EA
Add to cart
  • Unique COMBI-Rotor for tubes and PCR strips
  • Double the capacity of traditional mini centrifuges
  • Near silent operations
  • Starts and stops with opening and closing of the lid
  • Fixed speed
  • Perfect for personal use
Specifications
Capacity12 x 1.5/2.0 mL Tubes, 32 x 0.2 mL Tubes, 4 x PCR Strips
Max. Speed5,500 rpm
Max. RCF2,000 x g
Height (English) Exterior4.4 in.
Height (Metric) Exterior11.2 cm
Length (English) Exterior5.5 in.
Length (Metric) Exterior14 cm
Width (English) Exterior5.5 in.
Width (Metric) Exterior14 cm
Certifications/ComplianceCE
IncludesCOMBI-Rotor for tubes and strips
Safety FeaturesOperation stops when lid is opened
Electrical Requirements100/240 V 50/60 Hz
Depth (English)7.9 in.
Depth (Metric)20 cm
Wattage50 w
Rotor IncludedYes
Warranty2 Years
RefrigeratedNo
Net Weight2.2 lb.
Unit SizeEA
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link
1x1 image pixel for data collection