Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Actual product may vary
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Fisherbrand
Samco™ Graduated Transfer Pipettes
Measure more than one volume accurately by using these graduated polyethylene transfer pipettes. Built-in pipette bulb eliminates need for plugging, packaging, and autoclaving.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)SterilityGraduationsPackaging Type
FKS153678133.9 mLNon-sterile0.25, 0.50, 0.75, 1 mLBulk 500/Pk 10Pk/Cs 5000/Cs
FIS13-711-243.9 mLNon-sterile0.25, 0.50, 0.75, 1 mLBulk 500/Pk 10Pk/Cs 5000/Cs
FKS123844687.5 mLNon-sterile0.5, 1.0, 1.5, 2.0, 2.5, 3.0 mL
212-20S3.9 mLSterile0.25, 0.50, 0.75, 1 mL20/Bag, 500/Pk., 4000/Cs.
FKS153078235.8 mLSterile0.25, 0.50, 0.75, 1 mLPack of 500
2257.7 mLNon-sterile1, 2, 3 mLBulk 500/Pk 10Pk/Cs 5000/Cs
225-1S7.7 mLSterile1, 2, 3 mLIndividually Wrapped 500/Pk 8Pk/Cs 4000/Cs
FIS22-030-3207.7 mLSterile1, 2, 3 mLIndividually Wrapped 500/Pk 8Pk/Cs 4000/Cs
FIS13-711-507.7 mLSterile1, 2, 3 mL20/Bag, 500/Pk., 4000/Cs.
FIS13-711-431.5 mLNon-sterile0.1 , 0.2, 0.3 mLBulk 500/Pk 8Pk/Cs 4000/Cs
FKS102917901.5 mLSterile0.1, 0.2, 0.3 mLIndividually Wrapped 500/Pk 8Pk/Cs 4000/Cs
FKS1233447813.7 mLNon-sterile1, 2, 3, 4, 5 mLBulk 250/Pk 10Pk/Cs 2500/Cs
Catalog number FKS15367813
Price (SGD)
78.00
PK
Add to cart
Volume (Metric):
3.9 mL
Sterility:
Non-sterile
Graduations:
0.25, 0.50, 0.75, 1 mL
Packaging Type:
Bulk 500/Pk 10Pk/Cs 5000/Cs
Price (SGD)
78.00
PK
Add to cart
Accurately measure more than one volume by using Thermo Scientific™ Samco™ Graduated Transfer Pipettes. Single-unit design features a built-in pipette bulb that eliminates need for plugging, packaging, and autoclaving while preventing cross-contamination.
  • Made of shatterproof, nontoxic polyethylene
  • Low-affinity surface prevents proteins binding in research applications
  • Can be frozen in liquid nitrogen, or heat sealed for storage
Specifications
Bulb Draw (Metric)2.1 mL
DescriptionTransfer Pipette, Small Bulb, Graduated to 0.25, 0.50, 0.75, 1mL
Diameter (Metric) Outer4.7 mm
MaterialLDPE
SterilityNon-sterile
Drops/mL25
GraduatedYes
Graduations0.25, 0.50, 0.75, 1 mL
Packaging TypeBulk 500/Pk 10Pk/Cs 5000/Cs
Volume (Metric)3.9 mL
Length (Metric)14.0 cm
TypeGraduated Transfer Pipet