1x1 image pixel for data collection
Nalgene™ Right-to-Understand Safety Wash Bottles featuring Globally Harmonized System (GHS) labeling
Nalgene™ Right-to-Understand Safety Wash Bottles featuring Globally Harmonized System (GHS) labeling
Actual product may vary
Fisherbrand
Nalgene™ Right-to-Understand Safety Wash Bottles featuring Globally Harmonized System (GHS) labeling
Prevent chemical accidents in the lab with these Wash Bottles. Bottles are labeled following the Globally Harmonized System (GHS).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberClosure ColorChemical Label
FKS15363358GreenMethanol
2428-0507NaturalDeionized Water
FIS13-006-159NaturalDeionized Water
FKS15343368NaturalDeionized Water
2428-0504YellowIsopropanol
FIS13-006-156YellowIsopropanol
2428-0505NaturalDistilled Water
2428-0503GreenMethanol
FIS13-006-155GreenMethanol
2428-0502WhiteEthanol
FIS13-006-154WhiteEthanol
FKS15333358WhiteEthanol
2428-0506WhiteSodium Hypochlorite
Catalog number FKS15363358
Price (SGD)
199.00
PK
Estimated availability date 01-Oct-2025
Add to cart
Closure Color:
Green
Chemical Label:
Methanol
Price (SGD)
199.00
PK
Add to cart
Prevent chemical accidents in the lab with Nalgene™ Right-to-Understand LDPE Wash Bottles. Bottles are pre-labeled with chemical identification and safe handling information following the Globally Harmonized System (GHS) specifications as applicable to secondary laboratory containers. Wash bottles are ready to use when you transfer common laboratory chemicals from manufacturers' containers. Self-venting wash bottles eliminate risk of bottles self-dispensing.
  • Bottles are made from high quality Nalgene materials for low extractables and long life in the lab
  • Printed with 100% Globally Harmonized System (GHS) chemical safety information.  No other system markings are used on the bottles, which eliminates confusion
  • Color coded for easy chemical  identification at a glance
  • Dispensing nozzles are carefully molded to deliver a straight stream for precision dispensing and safety
  • Self-venting, so contents stay in bottle with no risk of leakage
  • Designed to dispense through the top with a closure and stem molded in one piece for uniform leakproof service
Right-to-Understand Safety Wash Bottles for acetone and other organic solvents are available in FEP (part number 2429-0501) for better chemical compatibility and product longevity.
Specifications
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorNatural
DescriptionWith stem and draw tube
MaterialLDPE
No. per Case24
No. per Pack6
Closure ColorGreen
GraduationsNo
Certifications/ComplianceCompliant with GHS Labeling Guidelines
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedNo
MouthNarrow
ShapeCylindrical
Product TypeSafety Wash Bottle
Chemical LabelMethanol
Closure MaterialPolypropylene
Closure Size28 mm
Closure TypeScrew Cap
Through the Closure DispensingYes
Unit SizePK
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link