Blocks for Fisherbrand™ Isotemp™ Digital Dry Baths/Block Heaters
Blocks for Fisherbrand™ Isotemp™ Digital Dry Baths/Block Heaters
Actual product may vary
Blocks for Fisherbrand™ Isotemp™ Digital Dry Baths/Block Heaters
Blocks for Fisherbrand™ Isotemp™ Digital Dry Baths/Block Heaters
Blocks for Fisherbrand™ Isotemp™ Digital Dry Baths/Block Heaters
Constructed of aluminum alloy
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)No. of Places
FKS153579382 mL28
FIS88-860-1031.5 mL28
Catalog number FKS15357938
Price (SGD)
361.00
EA
Add to cart
Capacity (Metric):
2 mL
No. of Places:
28
Price (SGD)
361.00
EA
Add to cart
A variety of blocks for any application
Specifications
Capacity (Metric)2 mL
For Use With (Equipment)Thermo Scientific Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
MaterialAluminum Alloy
No. of Places28
TypeBlock
Unit SizeEA