Isotemp™ Hot Plate, Ambient to 540°C, Ceramic
Isotemp™  Hot Plate, Ambient to 540°C, Ceramic
Actual product may vary
Fisherbrand
Isotemp™ Hot Plate, Ambient to 540°C, Ceramic
Shape: Square; Size: 4.25 x 4.25 in.; Plug Type: UK/AUS/CN-Style
Have Questions?
Catalog NumberDimensions (L x W x H)
FKS1535350810 x 5 x 3.6 in. (25.4 x 12.7 x 9.1 cm)
Catalog number FKS15353508
Price (SGD)
-
Dimensions (L x W x H):
10 x 5 x 3.6 in. (25.4 x 12.7 x 9.1 cm)
Experience exceptional safety and temperature performance for your routine protocols. Available in three sizes, providing flexibility from microscale chemistries to production operations.
Advanced technology
  • Strong magnetic coupling
  • Temperature adjustable in 1 degree increments
  • HOT TOP warning system designed to protect from accidental burns with prominent display when heating surface is above 50°C (122°F)
  • Automatic over-temperature protection
Flexible, robust design
  • Top plate choices in ceramic or aluminum
  • Raised display design protects electronics from spills
  • Optional temperature probe and splash guard protection shield
  • Variable sizes for different sample volumes
Specifications
DescriptionHotplate
Top Plate MaterialCeramic
Size4.25 x 4.25 in.
Temperature (Metric)Ambient to 540°C
Height (English)3.6 in.
Length (English)10 in.
Width (English)5 in.
Height (Metric)9.1 cm
Length (Metric)25.4 cm
Width (Metric)12.7 cm
Electrical Requirements230 V, 50/60 Hz
Frequency50/60 Hz
Plug TypeUK-Style, AUS-Style, CN-Style
ShapeSquare
Voltage230 V
TypeHot Plate
Certifications/ComplianceCE, ROHS Compliant
Warranty3 Years
Dimensions (L x W x H)10 x 5 x 3.6 in. (25.4 x 12.7 x 9.1 cm)
Unit SizeEA