Klercide Sporicidal Active Chlorine
Product Image
Fisherbrand
Klercide Sporicidal Active Chlorine
Deisgned to control all types of microbial contamination in cleanrooms. Ecolab™ Klercide Sporicidal Active Chloring is an effective biocide based on active chlorine, available in a ready to use spray bottle.
Have Questions?
Catalog NumberVolume (Metric)
FKS153524281 L
Catalog number FKS15352428
Price (SGD)
-
Volume (Metric):
1 L
  • Sterile by gamma irradiation
  • EN 1276 Bactericidal; EN 1650 Fungicidal (yeast); EN 13697 Bactericidal and fungicidal
  • Cleanroom processed
  • Unique Sterishield Delivery System ensures the sterility of the contents is preserved throughout use.
  • Available with complete supporting documentation
Specifications
Product TypeBiocide
SterilitySterile
AppearanceColorless
Certifications/ComplianceBactericidal EN 1276; Fungicidal EN 1650; Bactericidal and fungicidal EN 12397; Sporicidal EN 13704; Virucidal EN 14476
Container TypeSpray Bottle
DescriptionKlercide sporicidal active chlorine; Sporicidal Active Chlorine
For Use With (Application)Bactericidal, fungicidal, sporicidal, virucidal agent
Volume (Metric)1 L
Unit SizePK