U-Shaped Cell Culture Flasks
U-Shaped Cell Culture Flasks
Actual product may vary
Fisherbrand
U-Shaped Cell Culture Flasks
Designed to improve usability while maintaining the same environment for cell growth as previous designs. Corning™ U-Shaped Cell Culture Flasks have an ergonomic shape that reduces the number of corners, improves cell scraping and allows the use of a larger pipet.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure TypeSurface Coating
FKS15350591275 mLVentedTissue Culture Treated
CRG430641U275 mLVentedTissue Culture Treated
FIS07-202-000275 mLVentedTissue Culture Treated
Catalog number FKS15350591
Price (SGD)
1,418.00
PK
Add to cart
Capacity (Metric):
275 mL
Closure Type:
Vented
Surface Coating:
Tissue Culture Treated
Price (SGD)
1,418.00
PK
Add to cart
  • Cap Style: Available with Vented, Phenolic, or Plug Seal cap
  • 75 cm2 cell growth area
  • Fewer corners for easier scraping
  • Easier grip with better access to the cap
  • Tissue culture-treated surface
  • Nonpyrogenic
  • Resin meets USP Class VI specifications
  • Sterile to SAL 10-6
  • 100% leak tested
  • BSE/TSE compliant
  • Canted neck
  • Molded-in graduations for volumetric measurement estimation
  • Optional pipet removal of liquid via the pipet divot
  • Access for up to 50mL pipets
  • Stable stacking design
  • USP Class VI polystyrene
  • Manufactured in compliance with cGMP standards
Products are for research use only. Not intended for use in diagnostic or therapeutic procedures.
Specifications
Capacity (English)9.2 oz.
Capacity (Metric)275 mL
MaterialPolystyrene
For Use With (Application)Cell culture
IncludesLot numbers for ease in traceability
Neck StyleCanted
ShapeU-shaped
Surface CoatingTissue Culture Treated
GraduatedYes
SterilitySterile
Closure TypeVented
Culture Area75 cm2
Unit SizePK