Heat Sealer with Cutter
Heat Sealer with Cutter
Actual product may vary
Fisherbrand
Heat Sealer with Cutter
Seal bags, sheets or tubing quickly and efficiently. Heat Sealer with Cutter features a safe heating element that is only hot when in use. Timing knob allows for adjustable sealing duration. 450 Watt unit (220/230 VAC) includes a replacement part kit. Available with a UK or European plug.
Have Questions?
Catalog NumberDescription
FKS15350360UK Plug
Catalog number FKS15350360
Price (SGD)
1,082.00
EA
Add to cart
Description:
UK Plug
Price (SGD)
1,082.00
EA
Add to cart
Seal bags, sheets or tubing quickly and efficiently. Fisherbrand™ Heat Sealer with Cutter features a safe heating element that is only hot when in use. Timing knob allows for adjustable sealing duration. 450 Watt unit (220/230 VAC) includes a replacement part kit. Available with a UK or European plug.
  • Seal polyethylene or polypropylene bags, sheets or flat tubing from 0.01 to 0.8 mm thick.
  • Sliding blade removes excess material 19 mm from the seal.
  • Heating element measures 310 mm long and is only hot when sealing.
  • Timing knob ensures safe sealing.
    • Simply turn knob to set time (1-8 seconds).
    • Press handle down.
    • Remove when light indicates cycle is complete.
  • Includes replacement part kit (additional replacement part kits can be ordered separately):
    • Two extra blades
    • Two extra heating elements
    • Two extra PTFE cloths
Specifications
TypeCommercial Pouch Sealer
Dimensions (D x W x H)446 x 86 x 166 mm (closed)
For Use With (Equipment)Polyethylene or polypropylene bags, sheets or flat tubing
IncludesReplacement part kit (two each of: blades, heating elements, PTFE cloths), 153 cm power cord
Seal TimeAdjustable, 1 to 8 seconds
DescriptionUK Plug
Unit SizeEA