Check Order Status
Blocks for Fisherbrand™ Isotemp™ Digital Dry Baths/Block Heaters-Mixed Blocks
Copy product name
Copy catalog number
Copy catalog number and product name
Copy link
Actual product may vary
Blocks for Fisherbrand™ Isotemp™ Digital Dry Baths/Block Heaters-Mixed Blocks
Constructed of aluminum alloy
Have Questions?
Contact Us
Catalog Number
No. of Places
FKS15347948
28
Catalog number
FKS15347948
Price (SGD)
506.00
EA
Add to cart
No. of Places:
28
Price (SGD)
506.00
EA
Add to cart
A variety of blocks for any application
Specifications
Capacity (Metric)
18 x 1.5 mL, 10 x 2 mL
For Use With (Equipment)
Thermo Scientific Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
Material
Aluminum Alloy
No. of Places
28
Type
Block
Includes
1.5 mL and 2.0 mL
Unit Size
EA
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate
Safety Data Sheets
Share catalog number, name or link