BB-200 Series Electric Bunsen Burner
BB-200 Series Electric Bunsen Burner
Actual product may vary
BB-200 Series Electric Bunsen Burner
BB-200 Series Electric Bunsen Burner provides clean and easy operation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
FKS15341874Electric Bunsen Burner with MC5 Controller
FKS11728402Bunsen Burner (Controller is not included)
Catalog number FKS15341874
Price (SGD)
4,061.00
EA
Add to cart
Includes:
Electric Bunsen Burner with MC5 Controller
Price (SGD)
4,061.00
EA
Add to cart
  • Top cowl deflects heat away from hands
  • Air circulation from vented housing keeps base cool to touch
  • Energy efficient—only consumes 400 W of power
This electric Bunsen burner combines the advantages of a regular gas burner with the clean and easy operation of a heating mantle. Corrosion-resistant stainless steel burner features a cone-shaped coiled resistance heater which directs radiant heat out from the cavity. Cooling vents circulate air to cool the base, making it possible to hold while in use. A detachable cowl also deflects heat from the medium surface.
Ideal for noncontact radiant heating of materials in test tubes, crucibles, small flasks, and beakers. Effective heating of a test tube sample should be done 25 mm above the top of the burner. Crucible heating may be achieved by removing the cowl.
Note: To regulate the power input and temperature of the Bunsen burner, a heating controller is required. Without a controller, the burner operates continuously at full heating power. The Bunsen burner can be purchased with or without the recommended controller.
Specifications
IncludesElectric Bunsen Burner with MC5 Controller
Product TypeMantle with Controller
For Use With (Application)Heating teat tubes, crucibles, beakers
Unit SizeEA