LaboPlast™ Bio Disposable Spoon
LaboPlast™ Bio Disposable Spoon
Actual product may vary
Fisherbrand
LaboPlast™ Bio Disposable Spoon
Produced in a Cleanroom Class 7 (10,000) and individually wrapped in bio-PE film for disposable use. Green PE bio-plastic has all the suitable properties of conventional PE but is manufactured from renewable raw materials.
Have Questions?
Catalog NumberCapacity (Metric)
FKS153163372.5 mL
Catalog number FKS15316337
Price (SGD)
240.00
PK
Add to cart
Capacity (Metric):
2.5 mL
Price (SGD)
240.00
PK
Add to cart
  • Green PE bio-plastic has all the suitable properties of conventional polyethylene but is manufactured from renewable raw materials and can be completely recycled
  • The scoop stands horizontally as a result of the special handle design and the slightly angled front edge and can therefore be put down without the sample material trickling out.
  • Product and packaging made of renewable raw materials
  • Bio-PE, white
  • Individually wrapped
  • Production and packaging according to Cleanroom Class 7
  • With EU foodstuffs and FDA approval
Specifications
MaterialPolyethylene
Capacity (Metric)2.5 mL
DisposableSingle-use
SterilitySterile
ColorWhite
For Use With (Application)Foodstuffs
Quantity100
Length (Metric)127 mm
Unit SizePK