Sharps Container with Slide Access Lid
Sharps Container with Slide Access Lid
Actual product may vary
Fisherbrand
Sharps Container with Slide Access Lid
Prevent direct contact with sharps and accidental needle sticks. Fisherbrand™ Sharps Container with Silde Access Lid provides safe disposal of syringes, needles and other small sharps.
Have Questions?
Catalog number FKS15290009
Price (SGD)
137.20
Special offer
Ends: 15-Dec-2025
382.00
Save 244.80 (64%)
PK
Add to cart
Price (SGD)
137.20
Special offer
Ends: 15-Dec-2025
382.00
Save 244.80 (64%)
PK
Add to cart
  • Stackable to reduce storage space before use
  • 1.0L capacity
  • Clear lid to view fill level
  • Slide access lid for temporary and final closures for safety and security
  • Built-in needle removal port
  • Compatible with the Droplet™ Blood Collection Tray
  • Measures 115 x 115 x 192mm
  • Pack of 18
Specifications
Capacity (Metric)1 L
Closure TypeSlide Access
Closure ColorTranslucent
ColorYellow
Dimensions (L x W x H)115 x 115 x 192 mm
Height (Metric)192 mm
Length (Metric)115 mm
Quantity18/Pk.
StackableYes
Width (Metric)115 mm
IncludesSlide access lid
Unit SizePK