1x1 image pixel for data collection
Bulb Pipettes, One Mark, Class AS, Certified, ISO 648, DIN 12691
Bulb Pipettes, One Mark, Class AS, Certified, ISO 648, DIN 12691
Actual product may vary
Fisherbrand
Bulb Pipettes, One Mark, Class AS, Certified, ISO 648, DIN 12691
Pipette with the highest precision. Fisherbrand™ one mark Class AS pipettes are made of soda lime glass. Colour coded with blue markings.
Have Questions?
Catalog NumberVolume (Metric)
FKS15287935100 mL
Catalog number FKS15287935
Price (SGD)
73.00
PK
Estimated availability date 01-Oct-2025
Add to cart
Volume (Metric):
100 mL
Price (SGD)
73.00
PK
Add to cart
Pipette with the highest precision. Fisherbrand one mark Class AS pipettes are made of soda lime glass. Colour coded with blue markings.
  • Mark one pipet for precision measurment
  • Type one pipet with zero mark at the top
  • Soda lime glass 
  • Batch Certificate of Compliance is downloadable 
Specifications
Certifications/ComplianceISO 648, DIN 12691
ClassAS
MaterialSoda Lime Glass
GraduatedYes
Quantity2 Pack
Volume (Metric)100 mL
TypePipet
Unit SizePK
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link
1x1 image pixel for data collection