ST-30C Series Rear-facing Stereo Microscope
Actual product may vary
Fisherbrand
ST-30C Series Rear-facing Stereo Microscope
Designed and developed for the learning environment. Fisherbrand™ ST-30C Series Rear-facing Stereo Microscope has a choice between the innovative cordless LED illumination or conventional tungsten lighting system.
Have Questions?
Catalog NumberModelLighting TypeElectrical Requirements
FKS15254878ST-30C-6LEDLEDCharger 100 - 240V/Rechargeable batteries
Catalog number FKS15254878
Price (SGD)
1,113.00
Each
Model:
ST-30C-6LED
Lighting Type:
LED
Electrical Requirements:
Charger 100 - 240V/Rechargeable batteries
Price (SGD)
1,113.00
Each
  • 45° Inclined rear-facing binocular head with diopter adjustment on left eyetube
  • Widefield eyepieces 10X/20mm
  • 2X/4X magnification changer
  • Objectives are parfocal, parcentric and achromatic. Magnification is changed by turning the turret 180° to the click stop
  • Heavy-duty rack and pinion focusing block with slip clutch
  • Supplied standard with 80mm stage plates (Frosted glass, black and white)
  • Model ST-30c-2L00: Pole-stand with incident and transmitted 12V/10W tungsten illumination
  • Model ST-30C-6LED: Pole-stand with incident and transmitted rechargeable LED illumination
Specifications
IncludesBlack, white and frosted stage plates, dust cover, charger and batteries
Design TypeOptical: Greenough
Magnification Power2X to 4X
Weight (English)5.9 lb.
Weight (Metric)2.7 kg
Dimensions (L x W x H)7.4 x 4.5 x 11.2 in. (19 x 11.5 x 28.5 cm)
Electrical RequirementsCharger 100 - 240V/Rechargeable batteries
Product TypeMicroscope
ModelST-30C-6LED
Head StyleBinocular, Rear-facing, 45° inclined
Lighting TypeLED
EyepieceWF 10X/20 mm
Quantity1 Each
Unit SizeEach