Absorption Quartz Semi Micro Rectangular Cells with PTFE Lid, w/PTFE lid
Actual product may vary
Fisherbrand
Absorption Quartz Semi Micro Rectangular Cells with PTFE Lid, w/PTFE lid
Fit standard spectrophotometer cell holders
Have Questions?
Catalog NumberQuantity
FKS152460334/Pk., 8 Pk./Cs.
Catalog number FKS15246033
Price (SGD)
1,480.00
Pack
Quantity:
4/Pk., 8 Pk./Cs.
Price (SGD)
1,480.00
Pack
Fit standard spectrophotometer cell holders
  • All cells are 12.5mm wide
  • 10mm path length rectangular cells
  • Reduce reagent use with Semimicro cells
  • Quartz windows for transmission from far ultraviolet through the infrared
Specifications
TypeSemi-micro Absorption Cell
DescriptionBottom thickness: 9mm
Capacity (Metric)1000 μL
MaterialQuartz
Closure MaterialPTFE
Width (Metric) Interior4mm
Quantity4/Pk., 8 Pk./Cs.
Path Length10 mm
Wavelength Range200 to 2500 nm
Height (Metric)45 mm
For Use With (Equipment)Standard Spectrophotometer Cell Holder
Width (Metric)12.5 mm
For Use With (Application)Transmission from far ultraviolet through the infrared
ShapeRectangle
Unit SizePack