High Level Surface Disinfectant
High Level Surface Disinfectant
Actual product may vary
Fisherbrand
High Level Surface Disinfectant
Combines enhanced active ingredient molecules with micelle cleaning technology to provide swift penetration of the cell walls to ensure rapid cell death. Chemgene™ High Level Surface Disinfectant ensures that cell death is achieved rather than just suspending activity.
Have Questions?
Catalog NumberVolume (Metric)
FKS151766865 L
Catalog number FKS15176686
Price (SGD)
2,045.00
EA
Add to cart
Volume (Metric):
5 L
Price (SGD)
2,045.00
EA
Add to cart
Combines enhanced active ingredient molecules with micelle cleaning technology to provide swift penetration of the cell walls to ensure rapid cell death. Chemgene™ High Level Surface Disinfectant ensures that cell death is achieved rather than just suspending activity.
  • Biodegradable
  • Non-corrosive
  • Long shelf life
  • Diluted concentrate lasts up to 6 months
  • Compliant with REACH regulations and the Biocidal Product directive
Specifications
AppearanceBlue
Quantity1 Ea.
Volume (Metric)5 L
Unit SizeEA