Tygon™ R-3603 Lab Tubing
Tygon™ R-3603 Lab Tubing
Actual product may vary
Fisherbrand
Tygon™ R-3603 Lab Tubing
Formulated for resistance to flex fatigue and abrasion and is less permeable than rubber tubing. Tygon™ R-3603 Lab Tubing handles virtually all inorganic chemicals found in the lab.
Have Questions?
Catalog NumberDiameter (Metric) InnerDiameter (Metric) Outer
FKS151764940.8 mm2.4 mm
Catalog number FKS15176494
Price (SGD)
219.00
EA
Add to cart
Diameter (Metric) Inner:
0.8 mm
Diameter (Metric) Outer:
2.4 mm
Price (SGD)
219.00
EA
Add to cart
  • Bio-based formulation delivers excellent performance
  • Lot-to-lot consistency for reproducible results
  • Non-toxic and non-contaminating
  • Smooth, polished inner wall
  • Slips easily over fittings and grips securely for simple lab set-ups
  • Contains no BPA or phthalates
  • Standard sizes available to hold full vacuum at room temperature
Typical Applications
  • General laboratory
  • Food and beverage
  • Biopharmaceutical
  • Analytical instruments
  • Peristaltic and vacuum pumps
  • Ideal for condensers, incubators, desiccators, gas lines and drain line
Specifications
Certifications/ComplianceFDA Compliant, USP Class VI, NSF 51
AutoclavableAutoclavable
Diameter (English) Inner0.03 in.
Diameter (Metric) Inner0.8 mm
MaterialPlastic
Diameter (English) Outer0.09 in.
Diameter (Metric) Outer2.4 mm
Overall Length (English)49.2 ft.
Overall Length (Metric)15 m
Thickness (English)0.01 in.
Thickness (Metric)0.2 mm
For Use With (Application)General laboratory, food and beverage, biopharmaceutical, analytical instruments
Min. Operating Temperature (Metric)74°C
Packaging Quantity1 Ea.
SterilitySterile
ColorTransparent
Hardness56 (Shore A, 15 sec)
FlexibilityFlexible
Maximum Pressure69 psi
Vacuum Level4.8 bar (at 21°C)
Max. Operating Temperature (Metric)74°C
Product TypeTubing
Unit SizeEA