Transport Boxes
Transport Boxes
Actual product may vary
Fisherbrand
Transport Boxes
For transporting biological and clinical samples, as well as instruments and products that you want to keep clean and dry under testing environmental conditions. Fisherbrand™ Transport box is made of polycarbonate and available in three colors for user/owner identification.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
FKS15165564Clear, Blue
FIS12-006-916Clear, Blue
Catalog number FKS15165564
Price (SGD)
478.00
EA
Add to cart
Color:
Clear, Blue
Price (SGD)
478.00
EA
Add to cart
  • Polycarbonate construction and available in three colors for user/owner identification
  • Silicone water-tight seal and three lid clasps provide secure closure
  • Carry handle folds neatly into body space allowing the units to be stacked
  • Lid opens a full 180°, allowing total access to contents and making it easier to clean
  • Accommodates 13 and 16mm tubes in a 72-place onerack™, or similar sized/brand of rack
  • Separators provided to create own compartment sizes, or use empty to transport larger products and equipment
  • Autoclavable
Specifications
Product TypeWaterproof Transport Box
ColorClear, Blue
MaterialPolycarbonate
Dimensions (L x W x H)38.6 x 20.8 x 17 cm (15.2 x 8.2 x 6.7 in.)
Height (English)6.7 in.
Height (Metric)17 cm
IncludesPack of 1 (or Case of 12)
Length (English)15.2 in.
Length (Metric)38.6 cm
Weight (English)3.02 lb.
Weight (Metric)1.36 kg
Width (English)8.2 in.
Width (Metric)20.8 cm
Unit SizeEA