ThermoMixer™ C, Basic Device without Thermoblock
ThermoMixer™ C, Basic Device without Thermoblock
Actual product may vary
ThermoMixer™ C, Basic Device without Thermoblock
Use this temperature control device for heating and cooling almost any of your lab vessels. The Eppendorf ThermoStat™ C is the ideal device to accurately set and maintain temperatures.
Have Questions?
Catalog NumberCapacity (Metric)
FKS151589535μL to 50 mL
Catalog number FKS15158953
Price (SGD)
18,611.00
EA
Add to cart
Capacity (Metric):
5μL to 50 mL
Price (SGD)
18,611.00
EA
Add to cart
ThermoMixer C, without SmartBlock heating block, UK plug
  • Mixing frequency up to 3, 000rpm (depending on the Smart Block used)
  • Superior mixing performance due to 2D Mix-Control
  • Anti-spill technology prevents lid wetting and cross contamination
  • Low noise level for stress-free work
  • Simple and intuitive operation using pre-defined program keys
  • Ideally suited for Plasmid/RNA/DNA purification, cDNA synthesis, enzymatic reactions, denaturation of DNA, RNA and proteins, bacterial growth, lysis reactions at 100°C
  • Available separately: SmartBlock™ for 24 reaction vessels, 1.5mL (max. 2, 000rpm)
Specifications
Capacity (Metric)5μL to 50 mL
Plug TypeGB Plug
Temperature Range (Metric)15°C to 100°C
ModelThermoMixer C
Speed300 to 3000 rpm
Electrical Requirements220 to 240 V, 50/60 Hz
Voltage220/240 V
Frequency50 to 60 Hz
Unit SizeEA