1x1 image pixel for data collection
Stainless Steel Stirring Shafts with Propellers
Product Image
Fisherbrand
Stainless Steel Stirring Shafts with Propellers
Excellent at producing axial flows in vessels from top to bottom. Fisherbrand™ Stainless Steel Stirring Shafts with Propellers are for use with overhead stirrer units.
Have Questions?
Catalog NumberSizeIncludes
FKS15143007457mmPropellor, 108mm
Catalog number FKS15143007
Price (SGD)
1,029.00
EA
Estimated availability date 01-Oct-2025
Add to cart
Size:
457mm
Includes:
Propellor, 108mm
Price (SGD)
1,029.00
EA
Add to cart
  • Available in a range of sizes
  • Resistant to corrosion
Specifications
IncludesPropellor, 108mm
Quantity1 Each
Size457mm
TypeStirring Shaft
Unit SizeEA
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link