Non-sterile Nylon Syringe Filter
Non-sterile Nylon Syringe Filter
Actual product may vary
Fisherbrand
Non-sterile Nylon Syringe Filter
Designed for daily routine analysis. Fisherbrand™ Non-sterile Nylon Syringe Filter features a nylon membrane in a polypropylene housing for filtering aggressive aqueous solutions from a standard syringe. 25mm diameter membranes available with 0.2 or 0.45μm pore sizes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPore Size
FKS151314990.45 μm
FKS151214990.2 μm
Catalog number FKS15131499
Price (SGD)
74.00
PK
Add to cart
Pore Size:
0.45 μm
Price (SGD)
74.00
PK
Add to cart
  • Designed for use with aggressive aqueous solutions
  • Ideal for routine filtration of aqueous and solvent/aqueous mixtures
  • Hydrophilic 25mm diameter nylon membrane
  • Sturdy, chemically resistant polypropylene housing
Specifications
ColorWhite
Material (Membrane)Nylon
Pore Size0.45 μm
SterilityNon-sterile
For Use With (Application)Filtration of aggressive aqueous and solvent/aqueous mixtures
Housing MaterialPolypropylene
Quantity50/Pk.
TypeSyringe Filter
Diameter (Metric)25 mm
Unit SizePK