Borosilicate Glass Filtration System
Borosilicate Glass Filtration System
Actual product may vary
Fisherbrand
Borosilicate Glass Filtration System
Ideal for microbiology and preparation of HPLC solvents
Have Questions?
Catalog NumberDiameter (Metric) Membrane
FKS1511528447 mm
Catalog number FKS15115284
Price (SGD)
1,772.00
PK
Add to cart
Diameter (Metric) Membrane:
47 mm
Price (SGD)
1,772.00
PK
Add to cart
    For use with 47mm diameter membranes
  • Borosilicate glass porosity grade 3 sintered disc to support membrane
  • Complete assembly comprises of four components : Funnel 300mL capacity, Support base with sintered disc and vacuum connection with 40/38 socket, 1000ml conical flask suitable for use under vacuum with 40/38 cone, Metal retaining clip
  • Fully autoclavable
  • Each component is also available to order separately
Specifications
Connectorsvacuum
MaterialBorosilicate Glass
Material (Membrane)Borosilicate Glass
For Use With (Application)Microbiology and preparation of HPLC solvents
Funnel MaterialBorosilicate Glass
No. per Pack1Pack
TypeFiltration System
Volume (Metric)1000 mL
Diameter (Metric) Membrane47 mm
Unit SizePK