ThermoMixer™ C Accessory: Smartblock™ Thermoblocks
ThermoMixer™ C Accessory: Smartblock™ Thermoblocks
Actual product may vary
Fisherbrand
ThermoMixer™ C Accessory: Smartblock™ Thermoblocks
Maintain accurate uniform temperatures with these thermoblocks. Eppendorf™ Smartblock™ Thermoblocks are compatible with Eppendorf ThermoMixer™ C, and support a broad range of applications.
Have Questions?
Catalog NumberHolds
FKS1352847024 x 2mL
Catalog number FKS13528470
Price (SGD)
3,861.00
EA
Add to cart
Holds:
24 x 2mL
Price (SGD)
3,861.00
EA
Add to cart
  • Exchangeable in just seconds
  • Automatic block recognition for safe handling
Specifications
For Use With (Equipment)Eppendorf ThermoMixer C
Holds24 x 2mL
TypeBlock
Wash/Waste Bottle Capacity2mL x 24 Reaction vessels
Unit SizeEA