Non-sterile Transfer Pipettes, Transparent LDPE
Non-sterile Transfer Pipettes, Transparent LDPE
Actual product may vary
Fisherbrand
Non-sterile Transfer Pipettes, Transparent LDPE
Prevent cross contamination. Fisherbrand™ Non-sterile Transfer Pipets, Transparent LDPE are made of shatterproof, flexible plastic and are designed to replace glass Pasteur pipets. Graduated and non-graduated pipets available in a range of sizes and styles, including jumbo, thin stem and extra long.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Drops/mLGraduatedTip Style
FKS134991081 mL30YesStandard
FKS134691181 mL30NoStandard
FKS134391181 mL30YesExtended
FKS134391083 mL25YesStandard
FKS134591084 mL50NoStandard
FKS134491087 mL20NoExtra Long
FKS1344911810 mL18NoJumbo
Catalog number FKS13499108
Price (SGD)
38.00
PK
Add to cart
Volume (Metric):
1 mL
Drops/mL:
30
Graduated:
Yes
Tip Style:
Standard
Price (SGD)
38.00
PK
Add to cart
  • Built-in bulb eliminates the need for plugging, packaging and autoclaving
  • Flexible stem bends to sample from narrow or small tubes
  • Low affinity surface reduces loss of cells and valuable proteins due to binding
  • Soft plastic pipets made of non-toxic low-density polyethylene (LDPE)
  • Non-sterile, transparent and fully recyclable
Specifications
MaterialLDPE
Drops/mL30
GraduatedYes
Graduations0.25 mL
Quantity500 Pack
Tip StyleStandard
Volume (Metric) Dispense33 μL drop
Volume (Metric)1 mL
Length (Metric)150 mm
TypeTransfer Pipet
Unit SizePK