Under Bench Cabinet for Acids and Alkalis SL-CLASSIC-UB
Under Bench Cabinet for Acids and Alkalis SL-CLASSIC-UB
Actual product may vary
Fisherbrand
Under Bench Cabinet for Acids and Alkalis SL-CLASSIC-UB
Safe and approved storage of aggressive, non-flammable hazardous materials in working areas.
Have Questions?
Catalog NumberDoor Style
FKS13479716Wing, Left-hinged
Catalog number FKS13479716
Price (SGD)
3,328.00
EA
Add to cart
Door Style:
Wing, Left-hinged
Price (SGD)
3,328.00
EA
Add to cart
Safe and approved storage of aggressive, non flammable hazardous materials in working areas
Function / construction
  • Robust construction and longevity: made out of highly resistant melamin resin special plates
  • User-friendly: access to all containers, 93° opening of the wing door, interior of the cabinet completely visible
  • No unauthorised use: door lockable with cylinder lock and locking state indicator (red/green)
  • Mobile: castors with plinth (optionally available), easy moving of the cabinet
  • Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system
Certificates
  • type tested in accordance with EN 14727
  • EN 16121/16122 conformity
  • GS certified
Specifications
Certifications/ComplianceDIN EN 16121/16122 conformity, GS certified
Configuration1 x pull-out shelf (panel melamine resin-coated/PP)
Depth (Metric) Exterior570 mm
Depth (Metric) Interior531 mm
DescriptionUnder bench cabinet for acids and alkalis SL-CLASSIC-UB model SL.060.059.UB.T light grey, interior equipment with 1 x pull-out shelf (panel/PP)
Dimensions (D x W x H)570 x 590 x 600 mm
Dimensions (D x W x H) Interior531 x 489 x 562 mm
Door StyleWing, Left-hinged
For Use With (Application)Safe and approved storage of aggressive, non fla mmable hazardous materials in working areas
Height (Metric) Exterior600 mm
Height (Metric) Interior562 mm
ModelSL.060.059.UB.T
No. of Doors1
No. of Shelves1
Shelf TypePull-out
Surface CoatingMelamine resin laminated
Width (Metric) Exterior590 mm
Width (Metric) Interior485 mm
ColorLight Gray
MaterialChipboard
Product TypeUnder bench cabinet for acids and alkalis
Unit SizeEA