Labeling Tape
Labeling Tape
Actual product may vary
Fisherbrand
Labeling Tape
Adheres to any clean surface, including Teflon™ fluoropolymer resin coated materials. Bel Art™ Labeling Tape peels off without leaving a sticky residue.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorDimensions (LxW)
FKS13465226Yellow13.7 m x 12.7 mm
FKS13415236Yellow13.7 m x 25.4 mm
Catalog number FKS13465226
Price (SGD)
251.00
PK
Add to cart
Color:
Yellow
Dimensions (LxW):
13.7 m x 12.7 mm
Price (SGD)
251.00
PK
Add to cart
  • Excellent for use with Write-On™ Multi-Roll Label Tape Dispensers (F13472-0000 and F13473-0000)
  • Pencil, ball-point pen or solvent ink marker remain legible under the most demanding laboratory conditions
  • Consists of colored paper with rubber resin adhesive that resists moisture and most solvents
  • Will withstand a maximum temperature of 157ºC (315ºF) for 30 minutes or 80ºC (176ºF) for 24 hours; can be used down to -80ºC (-112ºF)
  • Withstands a steam autoclaving cycle at 121ºC (250ºF) and gamma irradiation between 13.0 and 45.0kGy
Specifications
AutoclavableAutoclavable
Adhesive TypeSelf-adhesive
ColorYellow
Product TypeLabel Tape
Quantity6/Pk.
Temperature Range (Metric)-80°C to +157°C
Dimensions (LxW)13.7 m x 12.7 mm
Length (Metric)13.7 m
Width (Metric)12.70 mm
Unit SizePK