Search Thermo Fisher Scientific
Catalog Number | Volume (Metric) | Drops/mL | Graduated | Tip Style |
---|---|---|---|---|
FKS13449118 | 10 mL | 18 | No | Jumbo |
FKS13469118 | 1 mL | 30 | No | Standard |
FKS13499108 | 1 mL | 30 | Yes | Standard |
FKS13439118 | 1 mL | 30 | Yes | Extended |
FKS13439108 | 3 mL | 25 | Yes | Standard |
FKS13459108 | 4 mL | 50 | No | Standard |
FKS13449108 | 7 mL | 20 | No | Extra Long |