Check Order Status
HDPE, Rectangular Carboy with Spigot
Actual product may vary
Fisherbrand
HDPE, Rectangular Carboy with Spigot
For Use With:
Ideal for storing and dispensing lab water and reagents;
Color:
White
Have Questions?
Change view
Catalog Number
Capacity (Metric)
FKS13408039
20 L
FIS03-007-643
9 L
FIS03-007-644
20 L
3 Options
Catalog number
FKS13408039
Price (SGD)
381.00
EA
Add to cart
Capacity (Metric):
20 L
Price (SGD)
381.00
EA
Add to cart
Carboys Ideal for storing and dispensing lab water and reagents.
Ideal for storing and dispensing lab water and reagents
Durable construction for long service life
Molded graduations for 1 gallon and 51 increments
Choice of materials for general use or autoclavability
Space-saving rectangular design
Wide mouth closure allows for easy filling and cleaning access
Carboys with spigots include a hose barb insert to accommodate -+ to 5/16-in. ID tubing.
Spigot spout without the adapter fits 5/8-in. ID tubing
Materials meet FDA requirements for food use and USP VI criteria (non-toxic)
Specifications
Autoclavable
Not Autoclavable
Capacity (English)
5.3 gal.
Capacity (Metric)
20 L
Certifications/Compliance
FDA, USP VI
Color
White
For Use With (Application)
Ideal for storing and dispensing lab water and reagents
Material
HDPE
Mouth Style
Wide
Product Type
Carboy with Spigot
Shape
Rectangular