Ministat 230 Refrigerated Heating Bath Circulator
Actual product may vary
Fisherbrand
Ministat 230 Refrigerated Heating Bath Circulator
The smallest cooling circulators in the world, which can be placed in a laboratory fume cupboard or inside technical plant. Ministats are small, but offer sufficient performance for control of photometers, refractometers, viscometers, distillation apparatus, reaction vessels, mini-plants.
Have Questions?
Catalog NumberHeight (Metric)
FKS13351539476 mm
Catalog number FKS13351539
Price (SGD)
54,658.00
Pack
Height (Metric):
476 mm
Price (SGD)
54,658.00
Pack
  • Refrigerated heating circulator bath
  • Pilot ONE controller with Plug and Play technology
  • Precise temperature control
  • 5.7 in. touchscreen and comfortable menu navigation
  • Overtemperature and low level protection
  • Safety class III/FL (DIN 12876)
  • Powerful pressure/suction pump
  • Programmer with calendar and clock functions
  • Five-point calibration
  • Pt100 sensor connection
  • Temperature range: -40°C to 200°C
  • Cooling capacity (20°C): 0.42kW
  • Cooling capacity (0°C): 0.38kW
  • Cooling capacity (-40°C): 0.05kW
  • Heating capacity: 1,6 - 2,1 kW
  • Pressure pump max.: 22L/min / 0.7bar
  • Suction pump max.: 16L/min / 0.4bar
  • Pump connection: M16 x 1 male
Specifications
Cooling Capacity100°C at 0.42 kW, 20°C at 0.42 kW, 0°C at 0.38°CkW, to 10°C at 0.33°CkW, to 20°C (at 0.25 kW)
Pump TypeCirculation
Temperature Range-40°C to +200°C
DisplayTFT
FeaturesTFT Touchscreen Display , E-grade Fuction, Integrated Programmer, USB Inferance, Ethernet Inferance, RS232 inferance, Plug and play control technology, Compatible with spycontrol,Easy control operating concept, Protection safety fuction, Natural refrigerant
Heating Capacity1.6 - 2.1 kW
ModelMinistat 230
Pump Capacity22 L / min; 0.7 bar Pressure, 16 L / min; 0.4 bar Suction
Safety Class3/FL
Temperature Stability±0.02°C
Warranty3 years for removable (Plug & Play) electronic controls, 2 years for refrigeration components, 1 year for mechanical and electrical components
Width (Metric)255 mm
Height (Metric)476 mm
Weight (Metric)37 kg
Voltage208/240 V