Sera-Mag™ Carboxylate modified Magnetic  Beads, hydrophobic
Actual product may vary
Fisherbrand
Sera-Mag™ Carboxylate modified Magnetic Beads, hydrophobic
Excellent for coupling oligos or proteins directly to beads. Their cauliflower-like structure gives Sera-Mag beads high binding capacity. The carboxyl groups on these single shell magnetite beads can be activated by standard EDC/EDAC chemistry to react with free amino groups, forming amide bonds.
Have Questions?
Catalog NumberQuantity
FKS1327358915 mL
Catalog number FKS13273589
Price (SGD)
-
Quantity:
15 mL
  • High binding capacity
  • 1.0μm diameter
  • High carboxyl acid content for binding for binding DNA and proteins
  • Hydrophobic bead based on crosslinking requirements for the encapsulation layer
Specifications
Concentration5% solids (50mg/mL)
For Use With (Application)DNA Sample Prep, Protein Purification
Quantity15 mL
Diameter (Metric)1 μm
Unit SizePack
Contents & Storage
2°C to 8°C