▸
Microscope Slides
▸
Glass Microscope Slides
▸
Glass Microscopic Slides with Ground Edges
Actual product may vary
Fisherbrand
Glass Microscopic Slides with Ground Edges
Use Glass Microscopic Slides with Ground Edges in the laboratory for a clear view of your slide material for analysis and diagnosis.
Have Questions?
Catalog Number
Quantity
FKS13192131
50 Pack
Catalog number
FKS13192131
Price (SGD)
11.00
Pack
Add to cart
Quantity:
50 Pack
Price (SGD)
11.00
Pack
Add to cart
Extra white glass
Washed and polished
90 degree ground edges
Polished coating
Specifications
Edges
90°, Ground
Frosted
Yes
Length (Metric)
76 mm
Thickness (Metric)
1 to 1.2 mm
Width (Metric)
26 mm
Color
White
Coating
Polished
Quantity
50 Pack