Sieve Shaker
Actual product may vary
Fisherbrand
Sieve Shaker
For accurate and reproducible sieving
Have Questions?
Catalog NumberVoltage
FKS13177913230 V
Catalog number FKS13177913
Price (SGD)
22,454.00
Each
Voltage:
230 V
Price (SGD)
22,454.00
Each
Fritsch sieve shakers satisfy all requirements for accurate and reproducible sieving analysis and perform automatic sieve analysis with the control and evaluation software Autosieve. The Analysette 3 Pro guarantees absolutely reproducible sieving results due to automatic amplitude control, programme selection, can be integrated into any quality management system according to DIN EN ISO 9000, and has an RS232 interface.
Sieves, clamping lid, interposed sieve pans and sieve pans with and without outlet are also available for 100mm and 8inch sieves.Test sieves, as well as accessories for micro precision sieving and for use of the Analysette 3 as micro mill (for grinding small sample quantities), are available on request.
For quantitative particle size analysis of dry samples and suspensions of all kinds, by dry and wet sieving with woven test sieves.
Specifications
Voltage230 V
Product TypeSieve Shaker
Frequency50 Hz
Unit SizeEach