Calypso Ultrapure Water Still
Calypso Ultrapure Water Still
Actual product may vary
Fisherbrand
Calypso Ultrapure Water Still
Provides many of the proven features of the high-performance Cyclon Still at an economical price. Fistreem International™ Calypso Ultrapure Water Still is available with 2L/hr or 4L/hr outputs and can be bench or wall mounted.
Have Questions?
Catalog NumberOutput Flow RateVoltageWattage
FKS131660204 L/hr.240 V3.0 kW
Catalog number FKS13166020
Price (SGD)
16,152.00
EA
Add to cart
Output Flow Rate:
4 L/hr.
Voltage:
240 V
Wattage:
3.0 kW
Price (SGD)
16,152.00
EA
Add to cart
  • Full featured.
  • Economical.
  • Available with 2L/hr or 4L/hr outputs.
  • Patented 'cyclonic' vapor trap for highest quality, pyrogen-free water.
  • Borosilicate glass construction.
  • Durable, silica-sheathed heating elements.
  • Can be connected directly to a mains tap supply, or a pre-treated water supply.
  • Automatic flow controller governs water consumption rate and allows unit to be used over a wide tap water pressure range.
  • In the pre-treated mode, purified water is fed directly to the boiler and tap water is used for cooling purposes only.
  • Supplied with a polyethylene level sensor that automatically shuts off power and input water whenever the reservoir is full, enabling safe, unattended operation.
  • Offered with an optional reservoir tank that allows a reserve of up to 30 liters storage capacity.
  • Polyethylene liner is fitted with a bacterial air filter, fast flow tap and contents sight tube and provides a convenient and compact collection system.
  • Automatic shut off is provided by still controls.
  • High temperature cut-off protects the heaters in the event of water failure while operator safety is assured through totally enclosed glassware.
  • Fitted with a boiler drain valve for easy cleaning. Cleaning solution is simply introduced into the boiler via the constant level device - no dismantling is necessary.
  • Bench or wall mountable and supplied with all necessary connection hoses and wall brackets.
Specifications
Dimensions (L x W x H)790 x 260 x 325 mm
Voltage240 V
Wattage3.0 kW
Inlet Pressure70 to 560 kPa 10 to 80 lbf.in-²
Output Flow Rate4 L/hr.
Unit SizeEA