UV Lamp
Product Image
Fisherbrand
UV Lamp
Rich, robust UV lamp that features UV light acting as a bactericide to break down and photo-oxidize organic contaminants to polar or ionized species.
Have Questions?
Catalog NumberFor Use With (Equipment)
FKS13135980Used as a bactericide and to break down and photo-oxidize organic contaminants to polar or ionized species
Catalog number FKS13135980
Price (SGD)
2,386.00
EA
Add to cart
For Use With (Equipment):
Used as a bactericide and to break down and photo-oxidize organic contaminants to polar or ionized species
Price (SGD)
2,386.00
EA
Add to cart
  • UV light is used as a bactericide and to break down and photo-oxidise organic contaminants to polar or ionized species
  • The UV bactericidal action damages DNA and RNA polymerase at low doses thereby preventing replication
Specifications
For Use With (Application)Used as a bactericide and to break down and photo-oxidize organic contaminats to polar or ionized species.
For Use With (Equipment)Used as a bactericide and to break down and photo-oxidize organic contaminants to polar or ionized species
Unit SizeEA