Terumo™ Eccentric Luer-Slip Syringe
Terumo™ Eccentric Luer-Slip Syringe
Actual product may vary
Fisher Catalogue
Terumo™ Eccentric Luer-Slip Syringe
Manufactured with polypropylene for superior thermal and chemical resistance. Bunzl™ Terumo™ Eccentric Luer-Slip Syringes have a unique bevel design to facilitate the penetration of the skin and minimize trauma.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Graduations
FKS1311479450 mL1 mL
FKS1150730220 mL1 mL
Catalog number FKS13114794
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
50 mL
Graduations:
1 mL
  • Clear polypropylene barrel and plunger; double contact ring
  • Integrated plunger stop prevents accidental pull-out
  • Sterile, Latex-free
  • Bold, precise scale markings
  • Complies to ISO 7886 (DIS 7886)
  • Individually wrapped in tamper evident peel packs
  • Supplied without needles
  • Suitable for gases or liquids
  • Eccentric luer lock
For needles, please reference our disposable needles range, ISO 7886 (DIS 7886): 13127634, 13117614, 13144724, 13187434, 12349219, 13104354.
For research use only, unless otherwise indicated.
Specifications
Certifications/ComplianceISO 7886 (DIS 7886)
MaterialPolypropylene
For Use With (Application)For research use, for gases or liquids
Product TypeEccentric Luer-slip Syringe
DisposableSingle-use
Graduations1 mL
Quantity25/Pk.
Volume (Metric)50 mL
Connection TypeEccentric Luer Slip