PolarPack™ Refrigerant Gel Packs
PolarPack™ Refrigerant Gel Packs
Actual product may vary
Fisherbrand
PolarPack™ Refrigerant Gel Packs
Economical and practical refrigerant pack choice for shipping temperature-sensitive material such as food products, pharmaceuticals, and more. Thermosafe™ PolarPack™ Refrigerant Gel Packs are enclosed in durable polyethylene film.
Have Questions?
Catalog NumberWeight (English)
FKS130793328 oz./Pack
Catalog number FKS13079332
Price (SGD)
168.00
PK
Add to cart
Weight (English):
8 oz./Pack
Price (SGD)
168.00
PK
Add to cart
  • Reliable non-mechanical refrigeration for the safe transport of food products, pharmaceuticals, and other medical products.
  • Multi-layered exterior minimizes surface condensation which protects product from moisture damage.
  • Outer film layer assures products arrive in top condition by absorbing condensation.
  • Phase change formulas ideal for products requiring frozen temperatures or extreme cold that may not tolerate dry ice or CO2.
  • Laminated, metalized film exterior provides radiant heat barrier, resists punctures.
  • Available in several sizes
  • Reusable
  • Non-toxic
Specifications
Quantity72 Pk./Cs.
Weight (English)8 oz./Pack
Dimensions (LxWxH)6.5 x 3.5 x 0.75 in.
Unit SizePK