Specifications
Certifications/ComplianceNIST, DKD/PTB, ANSI/SAMA Z236.1-1983, ASTM, ISO/IEC 17025:2005, A2LA
For Use With (Application)Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Immersion TypeTotal
Length (English) Overall7.9 in.
Length (Metric)200 mm
Length (Metric) Overall200 mm
MaterialGlass
Product TypeThermometer
Max. Temperature (Metric)150°C
Min. Temperature (Metric)-10°C
Temperature Range (Metric)-10°C to +150°C
CalibratedYes
Accuracy±1°C, ±1.5°C above 110°C
Subdivision1°C
Unit SizeEach