Easy-Read™ Individually Calibrated Liquid-In-Glass Thermometers: Partial Immersion
Easy-Read™ Individually Calibrated Liquid-In-Glass Thermometers: Partial Immersion
Actual product may vary
Fisherbrand
Easy-Read™ Individually Calibrated Liquid-In-Glass Thermometers: Partial Immersion
Individual Calibration Report as needed for many standard operating procedures
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTemperature Range (Metric)Calibration Points
FKS13069142-20°C to +150°C0, 60, 120°C
FIS13-201-477-20°C to +110°C0, 4, 20, 37, 44.5, 103°C
Catalog number FKS13069142
Price (SGD)
1,383.00
EA
Add to cart
Temperature Range (Metric):
-20°C to +150°C
Calibration Points:
0, 60, 120°C
Price (SGD)
1,383.00
EA
Add to cart
  • Fill: Black spirit
  • Each thermometer is individually calibrated
  • Celsius-scale
  • Designed to be very easy to read
  • Replace mercury thermometers with the same accuracy without any hazards
  • Nontoxic, nonhazardous, biodegradable
  • NIST-Certified detailed, laminated report of calibration
  • Each thermometer features a black liquid and yellow back glass which creates a distinct contrast making this thermometer very easy to read
  • Individually calibrated to NIST standards at specific temperature and supplied with NIST report of calibration stating corrections to one-tenth the smallest division
  • Contain an environmentally friendly non-mercury liquid that is non-toxic, non-hazardous, and fully biodegradable
  • In case of breakage the liquid can be cleaned up easily with soap and water
  • “Lead-free”, yellow back glass and black liquid provides easy readability and fewer reading errors
  • Contains non-toxic, biodegradable, Enviro-Safe™ liquid
  • EnviroKleen™ certified
Applications:
Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Specifications
Calibration Points0, 60, 120°C
Certifications/ComplianceEach thermometer includes an individualized Thermometer Calibration Report stating corrections to one-tenth of the smallest division, unique report number and NIST standards used for the calibration. Individually serialized and made in the USA
For Use With (Application)Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
GradePrecision
Immersion TypePartial Immersion
Length (English) Overall12 in.
Length (Metric) Overall300 mm
MaterialGlass
Thermometer FillEnviro-Safe™ Liquid
Max. Temperature (Metric)150°C
Min. Temperature (Metric)-20°C
Temperature Range (Metric)-20°C to +150°C
Accuracy±1°C
Subdivision1°C
Immersion Length76 mm
Unit SizeEA