Dual Timer and Clock
Dual Timer and Clock
Actual product may vary
Fisherbrand
Dual Timer and Clock
Counts up and counts down. Brannan™ Dual Timer and Clock includes 12 or 24 hour clock display, stand, clip and magnetic fixing. Supplied with 1 AAA battery.
Have Questions?
Catalog number FKS13063023
Price (SGD)
-
  • 23hr, 59min, 59s count down and up with memory function
  • 12 or 24 hour clock display
  • Stand, clip and magnetic fixing
  • Dimensions: 72mm x 65mm x 27mm
  • LCD size: 42mm x 20mm
  • Supplied with 1 x AAA (LR03) battery
Specifications
Product TypeDual Timer
Time Range23 hr. 59 min. 59 sec.
DisplayDigital
Battery TypeAAA
Display Dimensions24 x 35 mm
Dimensions (L x W x H)68 x 53 x 14 mm
Length (Metric)68 mm
Width (Metric)14 mm
Height (Metric)53 mm
Weight (Metric)74 g
IncludesStand, clip and magnetic fixing
Unit SizeEA