PEEK Chromatography Tubing
PEEK Chromatography Tubing
Actual product may vary
Fisherbrand
PEEK Chromatography Tubing
An excellent replacement for stainless steel tubing
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (English) InnerOverall Length (English)Maximum PressureColor
FKS130298920.03 in.5 ft.7000 psi (483 bar)Green
FIS05-701-120.01 in.50 ft.7000 psi (483 bar)Blue
FIS05-701-190.005 in.100 ft.7000 psiNatural
Catalog number FKS13029892
Price (SGD)
301.00
EA
Add to cart
Diameter (English) Inner:
0.03 in.
Overall Length (English):
5 ft.
Maximum Pressure:
7000 psi (483 bar)
Color:
Green
Price (SGD)
301.00
EA
Add to cart
An excellent replacement for stainless steel tubing
Can be used in place of stainless steel tubing in most liquid analytical systems
  • Biocompatible, inert and easy to cut
  • Great for high-pressure applications
  • Wide choice of outer and inner diameters
  • Many sizes color-coded for easier inner diameter identification
Specifications
TypeTubing
ColorGreen
For Use With (Application)High-Pressure Applications
Diameter (English) Inner0.03 in.
Overall Length (English)5 ft.
MaterialPEEK
Maximum Pressure7000 psi (483 bar)
Unit SizeEA