Spirit-filled Max Min Thermometer
Spirit-filled Max Min Thermometer
Actual product may vary
Fisher Catalogue
Spirit-filled Max Min Thermometer
Measures maximum and minimum temperaturess with non-toxic spirit filling. Brannan™ Spirit-filled Max Min Thermometer can be wall-mounted and resets with the push of a button.
Have Questions?
Catalog NumberTemperature Range (Metric)
FKS13029212-30°C to +50°C
Catalog number FKS13029212
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Temperature Range (Metric):
-30°C to +50°C
Measures maximum and minimum temperatures with non-toxic spirit filling.
  • Wall mounting in plastic case
  • 205mm long.
  • Push button reset.
Specifications
Length (Metric)205 mm
Thermometer FillNon-toxic Spirit
Max. Temperature (Metric)50°C
Min. Temperature (Metric)-30°C
Temperature Range (Metric)-30°C to +50°C
Unit SizeEA