Eutech™ CyberScan PCD 650 Multi-Parameter Meter
Eutech™ CyberScan PCD 650 Multi-Parameter Meter
Actual product may vary
Fisherbrand
Eutech™ CyberScan PCD 650 Multi-Parameter Meter
Measure multiple parameters in a cost-effective and convenient way by using only one Eutech™ multi-parameter meter.
Have Questions?
Catalog NumberMeasurement Modes
FKS12991758Acidity, Alkalinity, Temperature, Salinity
Catalog number FKS12991758
Price (SGD)
11,568.00
EA
Add to cart
Measurement Modes:
Acidity, Alkalinity, Temperature, Salinity
Price (SGD)
11,568.00
EA
Add to cart
Combining the strengths of all CyberScan 600 series meters, the CyberScan PCD650 is a top-of-the-line waterproof handheld that enables concurrent measurement of pH, conductivity, dissolved oxygen and temperature, all at the same time. Simultaneously measures and displays four parameters. For use in industrial, environmental, water quality testing, an aquaculture applications.
Features:
  • ±1% full scale accuracy at 3-decimal resolution across a widemeasurement range
  • Ion-measurement capabilities with 3-digit resolution
  • Accepts two-cell and four-cell conductivity probe tomeasure up to 500m/s —metermeasures pure water
  • Selectablemulti-point calibration formore accurate conductivitymeasurements or single-point calibration for quick, convenient calibration with nomistakes
  • measures 90.00mg/L in DO concentration and 600% in DO saturation—one of the widest dissolved oxygenmeasurement ranges offered in the handheldmarket today
  • Fast and easy DO calibration with 100% calibration option against atmospheric air or 0% calibration against a zero solution
  • Salinity, Temperature and Pressure Compensation—accurate readings under varying conditions
  • No warm up time required with galvanic system ofmeasurement
  • One-Glance-Tells-All Screen Display with Backlight
  • Electrode status report, calibration data and simultaneous display of up to 4 parameters
  • Rugged and IP67-rated waterproof for applications in harsh environments.meter floats on water!
  • Easy-to-understand promptmessages for easy set-up, calibration and trouble-shooting
  • Recorded readings individually stamped with time and date in GLP compliant format
  • Auto-loggingmeter saves readings automatically within seconds to prevent data loss
  • HI-LO limit alarm for easy quality control –meter alerts when readings fall outside selected ranges
  • Wireless Information Technology: No wires, no cables—Transfer data frommeter to computer with the press of a button
  • Non-Volatilememory protects information andmeter settings, even when batteries run out
  • Four Levels of Password Protection prevents tampering or accidental data deletion
  • Calibration Alarm: Nomore out-dated calibration settings
Specifications
  • pH Range: -2.000 to 20.000 pH
  • of pH Calibration Points: 1 (Offset) to 6 points
  • pH Buffer Options: USA, NIST, DIN, PWB, Custom
  • mV Range: ±2000.0mV
  • Ion Concentration Range: 001 to 19900
  • DO Range: 0 to 90.00mg/L or ppm / 0 to 600%
  • Salinity Correction: Automatic
  • Barometric Pressure Correction: Automatic correction with built-in sensormmHg or 0.1 kPa
  • Conductivity Range.. to 500mS/cm
  • Cell Constant: 010 to 10.000
  • Temperature Coefficient: Linear & Pure
  • TDS Range: .. to 500.0 ppt
  • Salinity Range 0 to 80 ppt
  • Temperature Range: -10.0 to 110.0 °C / 14.0 to 230.0 °F (meter)
  • Temperature Compensation: Automatic /manual (0 to 100 °C)
  • memory: 500 Data Sets with Date/Time stamping & Security password protection
  • Output: IrDA Wireless Communications
  • Display:multi-data Back-lit dotmatrix; 7.2 cm (H) X 5.4 cm (W)
  • Special Functions: Auto-Calibration; Data-logging options with time/date indexed, GLP compliance; Simultaneous Display andmeasurement of 4 parameters; Cal-due alarm; pH slope/offset Display & Self-diagnostic
  • Power: 4 X 1.5 V 'AA' Alkor
Specifications
OutputsIrDA Wireless Communication
DescriptionWaterproof CyberScan PCD 650 pH/ mV/ Ion/ Conductivity/ TDS/ Resistivity/ Salinity/ Dissolved Oxygen Meter with double junction pH electrode ECFC7252203B, 4-cell Conductivity probe CONSEN923J, Dissolved Oxygen probe ECDOHANDYNEW, DAS software, multi-probe holder, power adapter & carrying kit set (all probes comes with 3m cable).
Display TypeLCD Graphic, Backlit
IP RatingIP67
Measurement ModesAcidity, Alkalinity, Temperature, Salinity
Range (Conductivity)Up to 199.9 μS/cm, 200 μS/cm to 1999 μS/cm, 2 mS/cm to 20 mS/cm
Range (pH)-2 to 20
Range (Salinity)0 to 80 ppt
Temperature Range (Metric)-10°C to +110°C
Unit SizeEA